5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L14185.MC
also known as L14185-MC
Price (USD)/ Each
45.20
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
45.20
Add to cart
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
Catalog numberL14185.MC
Price (USD)/ Each
45.20
-
Add to cart
Chemical Identifiers
CAS491-67-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)263.0-273.0?C
Appearance (Color)Yellow to green to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only