Thermo Scientific Chemicals

Diethoxymethylvinylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L14224.14
25 g, Each
Thermo Scientific Chemicals

Diethoxymethylvinylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L14224.14
25 g, Each
Quantity
Catalog number: L14224.14
also known as L14224-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5507-44-8
IUPAC Name
ethenyldiethoxymethylsilane
Molecular Formula
C7H16O2Si
InChI Key
MBGQQKKTDDNCSG-UHFFFAOYSA-N
SMILES
CCO[Si](C)(OCC)C=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text