Thermo Scientific Chemicals

Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals

Catalog number: L14254.18
50 g, Each
Thermo Scientific Chemicals

Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals

Catalog number: L14254.18
50 g, Each
Quantity
Catalog number: L14254.18
also known as L14254-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9085-42-1
MDL Number
MFCD00132715
Appearance (Color)
White to pale brown
Matrix
2% cross-linked Styrene-divinylbenzene
Particle Size
>300 micron : ≤10.0%
Form
Beads
Matrix
Trimethylbenzylammonium active functional group

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text