Thermo Scientific Chemicals

Dowex™ 1X8 200-400 (Cl), Thermo Scientific Chemicals

Catalog number: L14258.18
50 g, Each
Thermo Scientific Chemicals

Dowex™ 1X8 200-400 (Cl), Thermo Scientific Chemicals

Catalog number: L14258.18
50 g, Each
Quantity
Catalog number: L14258.18
also known as L14258-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12627-85-9
SMILES
*
MDL Number
MFCD00132718
Appearance (Color)
White to cream or yellow to orange to brown or black
Moisture holding capacity
≥39 to ≤45%
Comment
Ionic form: Cl-
Particle Size
-325mesh: ≤15%
Exchange capacity
Total exchange capacity: ≥1.2 meq/ml

Description

Dowex™ 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text