Thermo Scientific Chemicals

DL-Lactic acid, 80-85% aq. soln., Thermo Scientific Chemicals

Catalog number: L14259.0B
1000 g, Each
Thermo Scientific Chemicals

DL-Lactic acid, 80-85% aq. soln., Thermo Scientific Chemicals

Catalog number: L14259.0B
1000 g, Each
Quantity
Catalog number: L14259.0B
also known as L14259-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-21-5
IUPAC Name
2-hydroxypropanoic acid
Molecular Formula
C3H6O3
InChI Key
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILES
CC(O)C(O)=O
Assay (Aqueous acid-base Titration)
80-85%
Form
Viscous liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Optical Rotation
0 ? 0.5? (c=1.5 in 2.5M NaOH)

Description

DL-Lactic acid is used to produce serial products or widely used in food, vintage, beverage, drugs, polymerization, textile, leather, tobacco, feed, plastic chemicals, pesticide, polymer solution and other industry. Lactic acid is also used as an acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text