N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.
Solubility
Soluble in water.
Notes
Air sensitive and hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Polowczyk, I.; Urbano, B. F.; Rivas, B. L.; Bryjak, M.; Kabay, N. Equilibrium and kinetic study of chromium sorption on resins with quaternary ammonium and N-methyl-d-glucamine groups. Chem. Eng. J. 2016, 284, 395-404.
Samatya, S.; Tuncel, S. A.; Kabay, N. Boron removal from RO permeate of geothermal water by monodisperse poly(vinylbenzyl chloride-co-divinylbenzene) beads containing N-methyl-d-glucamine. Desalination 2015, 364, 75-81.