Thermo Scientific Chemicals

1,2-Diethoxyethane, 98%, Thermo Scientific Chemicals

Catalog number: L14283.AP
500 mL, Each
Thermo Scientific Chemicals

1,2-Diethoxyethane, 98%, Thermo Scientific Chemicals

Catalog number: L14283.AP
500 mL, Each
Quantity
Catalog number: L14283.AP
also known as L14283-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
629-14-1
IUPAC Name
1,2-diethoxyethane
Molecular Formula
C6H14O2
InChI Key
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
SMILES
CCOCCOCC
Form
Liquid
Assay (GC)
≥97.5%
Free acid (titration)
(as acetic acid) <0.15%
Water Content (Karl Fischer Titration)
<1.0%
Refractive Index
1.3900-1.3940 @ 20?C

Description

Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

Solubility
Soluble in water 34 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text