Thermo Scientific Chemicals

Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals

Catalog number: L14285.0B
1000 g, Each
Thermo Scientific Chemicals

Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals

Catalog number: L14285.0B
1000 g, Each
Quantity
Catalog number: L14285.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78922-04-0
IUPAC Name
3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular Formula
C13H10ClNO4S
InChI Key
APBOVLPLJFJSRI-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1
Exchange capacity
Exchange capacity: ≥1.8eq/L (H+)
Form
Beads
Moisture holding capacity
48-58% (H+)
Particle Size
≤4.0% (>1180 µm)
Appearance (Color)
Amber to pale brown or yellow-brown

Description

Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text