Thermo Scientific Chemicals

Triphenyltin chloride, 95%, Thermo Scientific Chemicals

Catalog number: L14289.36
500 g, Each
Thermo Scientific Chemicals

Triphenyltin chloride, 95%, Thermo Scientific Chemicals

Catalog number: L14289.36
500 g, Each
Quantity
Catalog number: L14289.36
also known as L14289-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
639-58-7
IUPAC Name
triphenyltin chloride
Molecular Formula
C18H17ClSn
InChI Key
ZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILES
Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystalline powder or powder
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%%
Comment
As a bulk commodity material may contain small quantites particulate matter from the manufacturing process
Melting Point (clear melt)
103-109°C

Description

Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (0.005g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text