Thermo Scientific Chemicals

(S)-(+)-Epichlorohydrin, 98+%, Thermo Scientific Chemicals

Catalog number: L14298.14
25 g, Each
Thermo Scientific Chemicals

(S)-(+)-Epichlorohydrin, 98+%, Thermo Scientific Chemicals

Catalog number: L14298.14
25 g, Each
Quantity
Catalog number: L14298.14
also known as L14298-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67843-74-7
IUPAC Name
(2S)-2-(chloromethyl)oxirane
Molecular Formula
C3H5ClO
InChI Key
BRLQWZUYTZBJKN-GSVOUGTGSA-N
SMILES
ClC[C@@H]1CO1
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless
Optical Rotation
+35 ? 2? (c=1 in methanol)
Identification (FTIR)
Conforms
Refractive Index
1.4355-1.4405 @ 20?C

Description

(S)-(+)-Epichlorohydrin is utilized for the synthesis of macquarimicins and hydroxyisoxazolidines and (+)-cis-sylvaticin, which is a potential antitumor agent. It is used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators. It acts as a solvent for natural and synthetic resins, gums, cellulose esters and ethers, paints, varnishes, nail enamels, lacquers and cement for celluloid. It is also used as a stabilizer and also useful as a bifunctional alkylating agent with the potential to form DNA cross-links.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Epichlorohydrin is utilized for the synthesis of macquarimicins and hydroxyisoxazolidines and (+)-cis-sylvaticin, which is a potential antitumor agent. It is used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators. It acts as a solvent for natural and synthetic resins, gums, cellulose esters and ethers, paints, varnishes, nail enamels, lacquers and cement for celluloid. It is also used as a stabilizer and also useful as a bifunctional alkylating agent with the potential to form DNA cross-links.

Solubility
Miscible with chloroform and methanol. Immiscible with water.

Notes
Hygroscopic. Store in a cool place. Incompatible with acids, bases, ammonia, amines, sodium and sodium oxides, zinc, magnesium, aluminum and its alloys and halides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text