1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L14302.06
also known as L14302-06
Price (USD)/ Each
61.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
61.80
Add to cart
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
Catalog numberL14302.06
Price (USD)/ Each
61.80
-
Add to cart
Chemical Identifiers
CAS6974-32-9
IUPAC Name[5-(acetyloxy)-3,4-bis(benzoyloxy)oxolan-2-yl]methyl benzoate
Molecular FormulaC28H24O9
InChI KeyGCZABPLTDYVJMP-UHFFFAOYNA-N
SMILESCC(=O)OC1OC(COC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Melting Point (clear melt)125.0-132.0?C
Optical Rotation43.5 ± 1.5? (C=0.5 in chloroform)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only