Thermo Scientific Chemicals

delta-Octanolactone, 97%, Thermo Scientific Chemicals

Catalog number: L14303.22
100 g, Each
Thermo Scientific Chemicals

delta-Octanolactone, 97%, Thermo Scientific Chemicals

Catalog number: L14303.22
100 g, Each
Quantity
Catalog number: L14303.22
also known as L14303-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
698-76-0
IUPAC Name
6-propyloxan-2-one
Molecular Formula
C8H14O2
InChI Key
FYTRVXSHONWYNE-UHFFFAOYNA-N
SMILES
CCCC1CCCC(=O)O1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4535-1.4585 @ 20?C

Description

delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text