Thermo Scientific Chemicals

Ethanolamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14322.22
100 g, Each
Thermo Scientific Chemicals

Ethanolamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14322.22
100 g, Each
Quantity
Catalog number: L14322.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2002-24-6
IUPAC Name
hydrogen 2-aminoethan-1-ol chloride
Molecular Formula
C2H8ClNO
InChI Key
PMUNIMVZCACZBB-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCO
Assay (Titration ex Chloride)
≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or lumps or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text