Thermo Scientific Chemicals

N-Fluoropyridinium trifluoromethanesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: L14324.MD
250 mg, Each
Thermo Scientific Chemicals

N-Fluoropyridinium trifluoromethanesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: L14324.MD
250 mg, Each
Quantity
Catalog number: L14324.MD
also known as L14324-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107263-95-6
IUPAC Name
1-fluoropyridin-1-ium trifluoromethanesulfonate
Molecular Formula
C6H5F4NO3S
InChI Key
JFZMMCYRTJBQQI-UHFFFAOYSA-M
SMILES
F[N+]1=CC=CC=C1.[O-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Melting Point (clear melt)
175-187?C

Description

N-Fluoropyridinium trifluoromethanesulfonate is used as a reagent for electrophilic fluorination and for a review of electrophilic N-F fluorinating agents. It is also used as a reactant for Pd-catalyzed arylation reactions and as a fluorination agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fluoropyridinium trifluoromethanesulfonate is used as a reagent for electrophilic fluorination and for a review of electrophilic N-F fluorinating agents. It is also used as a reactant for Pd-catalyzed arylation reactions and as a fluorination agent.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text