Thermo Scientific Chemicals

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14325.MF
50 mg, Each
Thermo Scientific Chemicals

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14325.MF
50 mg, Each
Quantity
Catalog number: L14325.MF
also known as L14325-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39637-99-5
IUPAC Name
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular Formula
C10H8ClF3O2
InChI Key
PAORVUMOXXAMPL-VIFPVBQESA-N
SMILES
CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
Assay (GC)
≥98.0%
Refractive Index
1.4675-1.4715 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

Solubility
Sparingly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text