Tetra-n-propylammonium hydroxide, 1M aq. soln., Thermo Scientific Chemicals
Tetra-n-propylammonium hydroxide, 1M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-propylammonium hydroxide, 1M aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L14328.14
also known as L14328-14
Price (USD)/ Each
47.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
47.90
Add to cart
Tetra-n-propylammonium hydroxide, 1M aq. soln., Thermo Scientific Chemicals
Catalog numberL14328.14
Price (USD)/ Each
47.90
-
Add to cart
Chemical Identifiers
CAS4499-86-9
IUPAC Nametetrapropylazanium hydroxide
Molecular FormulaC12H29NO
InChI KeyLPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES[OH-].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Refractive Index1.3710-1.3750 @ 20?C
Assay (Aqueous acid-base Titration)0.98-1.02M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only