Thermo Scientific Chemicals

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14332.MF
50 mg, Each
Thermo Scientific Chemicals

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L14332.MF
50 mg, Each
Quantity
Catalog number: L14332.MF
also known as L14332-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20445-33-4
IUPAC Name
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular Formula
C10H8ClF3O2
InChI Key
PAORVUMOXXAMPL-SECBINFHSA-N
SMILES
CO[C@@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.4675-1.4715 @ 20?C
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms

Description

(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text