Thermo Scientific Chemicals

Chromium(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: L14342.14
25 g, Each
Thermo Scientific Chemicals

Chromium(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: L14342.14
25 g, Each
Quantity
Catalog number: L14342.14
also known as L14342-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10049-05-5
IUPAC Name
λ²-chromium(2+) dichloride
Molecular Formula
Cl2Cr
InChI Key
XBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cr++]
Form
powder
Assay (unspecified)
>96.0%
Appearance (Color)
Green to grey

Description

Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, water and active metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text