Thermo Scientific Chemicals

D-Glucuronic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L14350.14
25 g, Each
Thermo Scientific Chemicals

D-Glucuronic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L14350.14
25 g, Each
Quantity
Catalog number: L14350.14
also known as L14350-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6556-12-3
IUPAC Name
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
Molecular Formula
C6H10O7
InChI Key
IAJILQKETJEXLJ-QTBDOELSSA-N
SMILES
O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Optical Rotation
35.0 ± 2.5? (C=6 in water)
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text