4-Hydroxyindole, 98%, Thermo Scientific Chemicals
4-Hydroxyindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxyindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L14352.06
also known as L14352-06
Price (USD)/ Each
70.65
Online exclusive
78.40 
Save 7.75 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
78.40 
Save 7.75 (10%)
Add to cart
4-Hydroxyindole, 98%, Thermo Scientific Chemicals
Catalog numberL14352.06
Price (USD)/ Each
70.65
Online exclusive
78.40 
Save 7.75 (10%)
-
Add to cart
Chemical Identifiers
CAS2380-94-1
IUPAC Name1H-indol-4-ol
Molecular FormulaC8H7NO
InChI KeyNLMQHXUGJIAKTH-UHFFFAOYSA-N
SMILESOC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green-grey to brown
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only