Mercaptoacetic acid sodium salt, 97%, Thermo Scientific Chemicals
Mercaptoacetic acid sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercaptoacetic acid sodium salt, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number L14356.36
also known as L14356-36
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
Add to cart
Mercaptoacetic acid sodium salt, 97%, Thermo Scientific Chemicals
Catalog numberL14356.36
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS367-51-1
IUPAC Namesodium 2-sulfanylacetate
Molecular FormulaC2H3NaO2S
InChI KeyGNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤4.5% (105?C/constant weight)
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or lumps
Mercaptoacetic acid sodium salt is used as a reagent that protects tryptophan in amino acid analysis. It is a commonly used reagent for bacteriological research to maintain reducing conditions in media. It also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercaptoacetic acid sodium salt is used as a reagent that protects tryptophan in amino acid analysis. It is a commonly used reagent for bacteriological research to maintain reducing conditions in media. It also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state.

Solubility
Soluble in water (1000g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Store under inert gas. It is hygroscopic in nature and sensitive to moisture.
RUO – Research Use Only

General References:

  1. Eiki Adachi. Three-Dimensional Self-Assembly of Gold Nanocolloids in Spheroids Due to Dialysis in the Presence of Sodium Mercaptoacetate. Langmuir. 2000, 16 (16), 6460-6463.
  2. Morton Klein, J. H. Brewer, J. E. Perez and Beatrice Day. The Inactivation of Influenza Virus by Mercurials and the Reactivation by Sodium Thioglycolate and Bal. The Journal of Immunology. 1948, 59 (2), 135-140.