Thermo Scientific™

Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol, Thermo Scientific Chemicals

Catalog number: L14357.18
50 g, Each
Thermo Scientific™

Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol, Thermo Scientific Chemicals

Catalog number: L14357.18
50 g, Each
Quantity
Catalog number: L14357.18
also known as L14357-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
760-93-0
Refractive Index
1.4505-1.4575 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥92.5%

Description

Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

Solubility
Miscible with ethanol and ether.

Notes
Moisture and light sensitive. Store in a cool place. Incompatible with acids, strong oxidizing agents, alcohols, amines and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text