Thermo Scientific Chemicals

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals

Catalog number: L14365.06
5 g, Each
Thermo Scientific Chemicals

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals

Catalog number: L14365.06
5 g, Each
Quantity
Catalog number: L14365.06
also known as L14365-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-30-1
IUPAC Name
6,13-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Molecular Formula
C14H4O6
InChI Key
YTVNOVQHSGMMOV-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=CC=C3C(=O)OC(=O)C4=CC=C1C2=C34
Appearance (Color)
White to cream to yellow to pale brown
Acid-base back titration
≥96.0 to ≤104.0%
Form
Powder
Identification (FTIR)
Conforms

Description

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text