Thermo Scientific Chemicals

trans-beta-Nitrostyrene, 97+%, Thermo Scientific Chemicals

Catalog number: L14367.06
5 g, Each
Thermo Scientific Chemicals

trans-beta-Nitrostyrene, 97+%, Thermo Scientific Chemicals

Catalog number: L14367.06
5 g, Each
Quantity
Catalog number: L14367.06
also known as L14367-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5153-67-3
IUPAC Name
[(1E)-2-nitroethenyl]benzene
Molecular Formula
C8H7NO2
InChI Key
PIAOLBVUVDXHHL-VOTSOKGWSA-N
SMILES
[O-][N+](=O)\C=C\C1=CC=CC=C1
Form
Crystals or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Yellow
Melting Point (clear melt)
53-61?C
Assay (GC)
≥97.0%

Description

trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text