Thermo Scientific Chemicals

(+)-S-Trityl-L-cysteine, 97%, Thermo Scientific Chemicals

Catalog number: L14384.06
5 g, Each
Thermo Scientific Chemicals

(+)-S-Trityl-L-cysteine, 97%, Thermo Scientific Chemicals

Catalog number: L14384.06
5 g, Each
Quantity
Catalog number: L14384.06
also known as L14384-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2799-07-7
IUPAC Name
2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Formula
C22H21NO2S
InChI Key
DLMYFMLKORXJPO-UHFFFAOYNA-N
SMILES
NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Melting Point
155-170°C (U.S. specification)
Proton NMR
Conform to structure (U.S. specification)
Optical Rotation
+113° ± 6° (c=0.8% in 0.04 M Ethanolic HCl) (U.S. specification)
Optical Rotation
+83.5 ± 2.0° (c=1 in acetic acid) (non-U.S. specification)
Solution Test
Clear solution (0.3g in 2ml AcOH) (U.S. specification)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text