Triethylamine trihydrofluoride, ca 37% HF, Thermo Scientific Chemicals
Triethylamine trihydrofluoride, ca 37% HF, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylamine trihydrofluoride, ca 37% HF, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14417.14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS73602-61-6
IUPAC Nametrihydrogen triethylamine trifluoride
Molecular FormulaC6H18F3N
InChI KeyIKGLACJFEHSFNN-UHFFFAOYSA-N
SMILES[H+].[H+].[H+].[F-].[F-].[F-].CCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA21.3-28.0% (w/w) 'free' HF content
Appearance (Color)Clear colorless to brown
Assay from Supplier's CofA32-43% total HF content
FormLiquid
Triethylamine trihydrofluoride acts as a mild and selective fluorinating agent used in the synthesis of acid fluorides and alkyl fluorides. It is also used in the synthesis of vicinal difluorides from epoxides, 3-fluoroazetidine. It is utilized as a selective reagent for the cleavage of O-silyl groups in carbohydrates, nucleotides and cyanohydrins. It is actively involved in the electrochemical fluorodeiodination of alkyl iodides and fluorodesilylation in beta-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylamine trihydrofluoride acts as a mild and selective fluorinating agent used in the synthesis of acid fluorides and alkyl fluorides. It is also used in the synthesis of vicinal difluorides from epoxides, 3-fluoroazetidine. It is utilized as a selective reagent for the cleavage of O-silyl groups in carbohydrates, nucleotides and cyanohydrins. It is actively involved in the electrochemical fluorodeiodination of alkyl iodides and fluorodesilylation in beta-lactams.

Solubility
Miscible with water, chloroform and hexanes.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and glass.
RUO – Research Use Only

General References:

  1. Convenient mild fluorinating agent which can be handled in glass equipment (long-term storage in glass is not recommended):
  2. Activated halides can be displaced to give the corresponding fluorides J. Fluorine Chem., 15, 423 (1980); 47, 467 (1990). Carbohydrate bromides or mesylates can be converted to fluorinated analogues with inversion of configuration: J. Fluorine Chem., 60, 49 (1993); Tetrahedron Lett., 31, 6527 (1990).
  3. In the presence of NBS, alkenes undergo trans-fluorobromination in high yield: Synthesis, 562 (1987); Liebigs Ann. Chem., 849 (1995); Org. Synth., 76, 159 (1998). Vinyl oxiranes undergo ring opening to give fluorohydrins: J. Fluorine Chem., 70, 1 (1995); in the presence of NBS, fluorobromination of the double bond occurs instead: J. Org. Chem., 59, 5277 (1994).
  4. A variety of compounds undergo electrochemical fluorination at activated positions. ɑ-Fluorinations of sulfides have been extensively studied by Fuchigami and others. The reaction usually requires the presence of an electron withdrawing group ɑ- to the sulfur atom, e.g. monofluorination of ethyl (phenylthio)acetate: J. Org. Chem., 56, 6731 (1991); 59, 5937 (1994); 60, 3459 (1995), and of phenylthio substituted nitrogen heterocycles including ß-lactams: Tetrahedron Lett., 33, 7017 (1992); J. Org. Chem., 57, 3755 (1992); 58, 4200 (1993).
  5. For electrochemical fluorodeiodination of alkyl iodides, see: Synlett, 999 (2000). Fluorodesilylation in ß-lactams has also been reported: J. Chem. Soc., Perkin 1, 1327 (1995).