Thermo Scientific Chemicals

Isobutyronitrile, 99%, Thermo Scientific Chemicals

Catalog number: L14470.AU
1000 mL, Each
Thermo Scientific Chemicals

Isobutyronitrile, 99%, Thermo Scientific Chemicals

Catalog number: L14470.AU
1000 mL, Each
Quantity
Catalog number: L14470.AU
also known as L14470-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-82-0
IUPAC Name
2-methylpropanenitrile
Molecular Formula
C4H7N
InChI Key
LRDFRRGEGBBSRN-UHFFFAOYSA-N
SMILES
CC(C)C#N
Infrared spectrum
Conforms
Form
Liquid
Refractive Index
1.3720-1.3755 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%

Description

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text