Trichloroethylene, 98%, Thermo Scientific Chemicals
Trichloroethylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroethylene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1000 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14474.AE
also known as L14474-AE
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
23.00 
Save 3.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
23.00 
Save 3.35 (15%)
Add to cart
Trichloroethylene, 98%, Thermo Scientific Chemicals
Catalog numberL14474.AE
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
23.00 
Save 3.35 (15%)
-
Add to cart
Chemical Identifiers
CAS79-01-6
IUPAC Name1,1,2-trichloroethene
Molecular FormulaC2HCl3
InChI KeyXSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILESClC=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4750-1.4790 @ 20?C
Assay (GC)> 97.5%
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

Solubility
Soluble in water (1g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light Sensitive to light. Handle and store under inert gas. Incompatible with bases and oxidizing agents.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. T M Vogel.; P L McCarty. Biotransformation of tetrachloroethylene to trichloroethylene, dichloroethylene, vinyl chloride, and carbon dioxide under methanogenic conditions. Appl. Environ. Microbiol. 1985, 49 (5), 1080-1083.
  2. D L Freedman.; J M Gossett. Biological reductive dechlorination of tetrachloroethylene and trichloroethylene to ethylene under methanogenic conditions. Appl. Environ. Microbiol. 1985, 55 (9), 2144-2151.