Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane
Solubility
Soluble in water (1g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light Sensitive to light. Handle and store under inert gas. Incompatible with bases and oxidizing agents.
RUO – Research Use Only
T M Vogel.; P L McCarty. Biotransformation of tetrachloroethylene to trichloroethylene, dichloroethylene, vinyl chloride, and carbon dioxide under methanogenic conditions. Appl. Environ. Microbiol. 1985, 49 (5), 1080-1083.
D L Freedman.; J M Gossett. Biological reductive dechlorination of tetrachloroethylene and trichloroethylene to ethylene under methanogenic conditions. Appl. Environ. Microbiol. 1985, 55 (9), 2144-2151.