Methyl acetate is used as a solvent in glues, paints and nail polish removers. It is also used in adhesives, sealants and coatings. Further, it finds use as solvent in chemical reactions as well as used for extraction purpose. It is involved in the production of acetic anhydride by carbonylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl acetate is used as a solvent in glues, paints and nail polish removers. It is also used in adhesives, sealants and coatings. Further, it finds use as solvent in chemical reactions as well as used for extraction purpose. It is involved in the production of acetic anhydride by carbonylation reaction.
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Yang, X.; Felsmann, D.; Kurimoto, N.; Krüger, J.; Wada, T.; Tan, T.; Carter, E. A.; Kohse-Höinghaus, K.; Ju, Y. Kinetic studies of methyl acetate pyrolysis and oxidation in a flow reactor and a low-pressure flat flame using molecular-beam mass spectrometry. Proc. Combust. Inst. 2015, 35 (1), 491-498.
Rasmussen, D. B.; Christensen, J. M.; Temel, B.; Studt, F.; Moses, P. G.; Rossmeisl, J.; Riisager, A.; Jensen, A. D. Ketene as a Reaction Intermediate in the Carbonylation of Dimethyl Ether to Methyl Acetate over Mordenite. Angew. Chem. Int. Ed. 2015, 54 (25), 7261-7264.
Nguyen, H. V. L.; Kleiner, I.; Shipman, S. T.; Mae, Y.; Hirose, K.; Hatanaka, S.; Kobayashi, K. Extension of the measurement, assignment, and fit of the rotational spectrum of the two-top molecule methyl acetate. J. Mol. Spectrosc. 2014, 299, 17-21.
Niza, N. M.; Tan, K. T.; Lee, K. T.; Ahmad, Z. Biodiesel production by non-catalytic supercritical methyl acetate: thermal stability study. Appl. Energy 2013, 101, 198-202.