Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals
Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14491.36
also known as L14491-36
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
Add to cart
Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals
Catalog numberL14491.36
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS818-08-6
IUPAC Namedibutylstannanone
Molecular FormulaC8H18OSn
InChI KeyJGFBRKRYDCGYKD-UHFFFAOYSA-N
SMILESCCCC[Sn](=O)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)47.0% - 48.0% Tin
Chloride content<0.07%
Comment<2.00% Monobutyltin
Loss on Drying<0.5%
Comment<2.8 Acetone Solubles
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only