Thermo Scientific Chemicals

Sodium L-lactate, 98+%, Thermo Scientific Chemicals

Catalog number: L14500.14
25 g, Each
Thermo Scientific Chemicals

Sodium L-lactate, 98+%, Thermo Scientific Chemicals

Catalog number: L14500.14
25 g, Each
Quantity
Catalog number: L14500.14
also known as L14500-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
867-56-1
IUPAC Name
sodium 2-hydroxypropanoate
Molecular Formula
C3H5NaO3
InChI Key
NGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES
[Na+].CC(O)C([O-])=O
Form
Crystals or powder or crystalline powder or lumps
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White
Assay from Supplier's CofA
≥98.0%
Identification (FTIR)
Conforms

Description

Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text