Thermo Scientific Chemicals

4-Benzyloxy-3,5-dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L14536.06
5 g, Each
Thermo Scientific Chemicals

4-Benzyloxy-3,5-dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L14536.06
5 g, Each
Quantity
Catalog number: L14536.06
also known as L14536-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97888-80-7
IUPAC Name
4-(benzyloxy)-3,5-dimethylbenzoate
Molecular Formula
C16H15O3
InChI Key
JABUPJCJZZNUFK-UHFFFAOYSA-M
SMILES
CC1=CC(=CC(C)=C1OCC1=CC=CC=C1)C([O-])=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale yellow to yellow to cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
155.5-161.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text