4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L14554.06
also known as L14554-06
Price (USD)/ Each
371.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
371.00
Add to cart
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberL14554.06
Price (USD)/ Each
371.00
-
Add to cart
Chemical Identifiers
CAS16261-80-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (non-U.S. specification)
Assay from Suppliers CofA≥96.0% (HPLC + Titration: U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jiang, S.; Xu, Q.; Zhu, M.; Yu, X.; Deng, W. Kinetic Resolution of α-Methylene- β-hydroxy Esters Catalyzed by Acyl Transfer Catalyst An-PIQ. J. Org. Chem. 2015, 80 (6), 3159-3169.
  2. Allu, S.; Swamy, K. C. K. Ruthenium-Catalyzed Oxidative Annulation of 6-Anilinopurines with Alkynes via C H Activation: Synthesis of Indole-Substituted Purines/Purine Nucleosides. Adv. Synth. Catal. 2015, 357 (12), 2665-2680.