Thermo Scientific Chemicals

Poly(methylhydrosiloxane), Thermo Scientific Chemicals

Catalog number: L14561.30
250 g, Each
Thermo Scientific Chemicals

Poly(methylhydrosiloxane), Thermo Scientific Chemicals

Catalog number: L14561.30
250 g, Each
Quantity
Catalog number: L14561.30
also known as L14561-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63148-57-2
Molecular Formula
(CH4OSi)n
InChI Key
QNWOFLWXQGHSRH-UHFFFAOYSA-N
SMILES
C[SiH](O[Si](C)(C)C)O[Si](C)(C)C
MDL Number
MFCD00084478
Appearance (Color)
Clear colorless
Comment
Hydride content 96-100 mole%
Viscosity
15-25 cSt (from supplier's Certificate of Analysis)
Identification (FTIR)
Conforms
Refractive Index
1.3960-1.4000 @ 20?C

Description

Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text