Thermo Scientific Chemicals

Cyclopentylacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L14562.06
5 g, Each
Thermo Scientific Chemicals

Cyclopentylacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L14562.06
5 g, Each
Quantity
Catalog number: L14562.06
also known as L14562-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1122-99-2
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Refractive Index
1.4595-1.4645 @ 20?C

Description

Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with bases and oxidizing agents. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text