2,2,7-Trimethyloctane-3,5-dione, 97%, Thermo Scientific Chemicals
2,2,7-Trimethyloctane-3,5-dione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,7-Trimethyloctane-3,5-dione, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L14639.06
also known as L14639-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
2,2,7-Trimethyloctane-3,5-dione, 97%, Thermo Scientific Chemicals
Catalog numberL14639.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS69725-37-7
IUPAC Name2,2,7-trimethyloctane-3,5-dione
Molecular FormulaC11H20O2
InChI KeyKEBPOWLGOOTMPK-UHFFFAOYSA-N
SMILESCC(C)CC(=O)CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4575-1.4615 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only