Thermo Scientific Chemicals

Benzyl L-lactate, 97%, Thermo Scientific Chemicals

Catalog number: L14645.14
25 g, Each
Thermo Scientific Chemicals

Benzyl L-lactate, 97%, Thermo Scientific Chemicals

Catalog number: L14645.14
25 g, Each
Quantity
Catalog number: L14645.14
also known as L14645-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56777-24-3
IUPAC Name
benzyl 2-hydroxypropanoate
Molecular Formula
C10H12O3
InChI Key
ZYTLPUIDJRKAAM-UHFFFAOYNA-N
SMILES
CC(O)C(=O)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.5100-1.5150 @ 20?C
Form
Liquid
Optical Rotation
-16.8 ± 0.5? (C=3 in methanol)

Description

Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text