Thermo Scientific Chemicals

Petroleum ether 40/60, Thermo Scientific Chemicals

Catalog number: L14649.0F
2500 mL, Each
Thermo Scientific Chemicals

Petroleum ether 40/60, Thermo Scientific Chemicals

Catalog number: L14649.0F
2500 mL, Each
Quantity
Catalog number: L14649.0F
also known as L14649-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8032-32-4
Refractive Index
1.3710-1.3750 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

Solubility
Insoluble in water.

Notes
Flammable. May be moisture sensitive.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text