Thermo Scientific Chemicals

1-Phenyl-2-pyrrolidinone, 99%, Thermo Scientific Chemicals

Catalog number: L14660.14
25 g, Each
Thermo Scientific Chemicals

1-Phenyl-2-pyrrolidinone, 99%, Thermo Scientific Chemicals

Catalog number: L14660.14
25 g, Each
Quantity
Catalog number: L14660.14
also known as L14660-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4641-57-0
IUPAC Name
1-phenylpyrrolidin-2-one
Molecular Formula
C10H11NO
InChI Key
JMVIVASFFKKFQK-UHFFFAOYSA-N
SMILES
O=C1CCCN1C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or granules or fused solid
Melting Point
62-69°C
Appearance (Color)
Colorless to white to pale cream or pale yellow or pale pink
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

Solubility
Soluble in chloroform and ethyl Acetate. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text