5-Isopropyl-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
5-Isopropyl-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Isopropyl-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number L14737.14
also known as L14737-14
Price (USD)/ Each
153.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
153.00
Add to cart
5-Isopropyl-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberL14737.14
Price (USD)/ Each
153.00
-
Add to cart
Chemical Identifiers
CAS85902-68-7
IUPAC Name2-methoxy-5-(propan-2-yl)benzaldehyde
Molecular FormulaC11H14O2
InChI KeyFPZHOBMVPBJNKL-UHFFFAOYSA-N
SMILESCOC1=C(C=O)C=C(C=C1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
Refractive Index1.5360-1.5410 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only