Thermo Scientific Chemicals

2,6-Difluorophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L14779.06
5 g, Each
Thermo Scientific Chemicals

2,6-Difluorophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L14779.06
5 g, Each
Quantity
Catalog number: L14779.06
also known as L14779-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65295-69-4
IUPAC Name
1,3-difluoro-2-isocyanatobenzene
Molecular Formula
C7H3F2NO
InChI Key
YXHDLKWTPVMIOH-UHFFFAOYSA-N
SMILES
FC1=CC=CC(F)=C1N=C=O
Refractive Index
1.4925-1.4975 @ 20?C
Appearance (Color)
Colorless to white to pale yellow
Form
Fused solid or clear liquid as melt
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text