3-Buten-2-ol, 97%, Thermo Scientific Chemicals
3-Buten-2-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Buten-2-ol, 97%, Thermo Scientific Chemicals

Important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L14787.14
also known as L14787-14
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
3-Buten-2-ol, 97%, Thermo Scientific Chemicals
Catalog numberL14787.14
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS598-32-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
FormLiquid
Refractive Index1.4115-1.4165 @ 20?C
Assay (GC)>96.0%
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only