Thermo Scientific Chemicals

4-Iodophenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L14806.03
1 g, Each
Thermo Scientific Chemicals

4-Iodophenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L14806.03
1 g, Each
Quantity
Catalog number: L14806.03
also known as L14806-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13116-27-3
IUPAC Name
(4-iodophenyl)hydrazine
Molecular Formula
C6H7IN2
InChI Key
IQMLCMKMSBMMGR-UHFFFAOYSA-N
SMILES
NNC1=CC=C(I)C=C1
Assay (HPLC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
94.0-107.0?C
Appearance (Color)
Yellow to dark brown to brown
Form
Crystals or powder or crystalline powder or fused solid

Description

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text