Thermo Scientific Chemicals

4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: L14810.06
5 g, Each
Thermo Scientific Chemicals

4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: L14810.06
5 g, Each
Quantity
Catalog number: L14810.06
also known as L14810-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93048-16-9
IUPAC Name
(4-methoxy-2-methylphenyl)hydrazine hydrochloride
Molecular Formula
C8H13ClN2O
InChI Key
OIFKWIPSJGNUCC-UHFFFAOYSA-N
SMILES
Cl.COC1=CC(C)=C(NN)C=C1
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Appearance (Color)
White to cream to pale brown or pink
Form
Powder or granules

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text