Thermo Scientific Chemicals

Hexafluorosilicic acid, 35% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L14829.36
500 g, Each
Thermo Scientific Chemicals

Hexafluorosilicic acid, 35% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L14829.36
500 g, Each
Quantity
Catalog number: L14829.36
also known as L14829-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16961-83-4
IUPAC Name
dihydrogen hexafluorosilanediuide
Molecular Formula
F6H2Si
InChI Key
OHORFAFFMDIQRR-UHFFFAOYSA-P
SMILES
[H+].[H+].F[Si--](F)(F)(F)(F)F
Appearance (Color)
Clear to slightly hazy, colorless to pale yellow
Form
Liquid
Assay (Aqueous acid-base Titration)
33-37% w/w

Description

Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.

Solubility
Miscible with water.

Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text