Thermo Scientific Chemicals

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%, Thermo Scientific Chemicals

Catalog number: L14835.06
5 g, Each
Thermo Scientific Chemicals

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%, Thermo Scientific Chemicals

Catalog number: L14835.06
5 g, Each
Quantity
Catalog number: L14835.06
also known as L14835-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
202823-67-4
IUPAC Name
3,6-dichloro-4,5-diazatricyclo[6.2.2.0²,⁷]dodeca-2(7),3,5-triene
Molecular Formula
C10H10Cl2N2
InChI Key
GZIFYLGNTOAIJJ-UHFFFAOYSA-N
SMILES
ClC1=NN=C(Cl)C2=C1C1CCC2CC1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
207.5-216.5?C

Description

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text