Thermo Scientific Chemicals

Polyphosphoric acid, ca 84% (as phosphorus pentoxide), Thermo Scientific Chemicals

Catalog number: L14856.AP
500 mL, Each
Thermo Scientific Chemicals

Polyphosphoric acid, ca 84% (as phosphorus pentoxide), Thermo Scientific Chemicals

Catalog number: L14856.AP
500 mL, Each
Quantity
Catalog number: L14856.AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8017-16-1
IUPAC Name
Polyphosphoric acid
Molecular Formula
(HO3P)n
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
[H]OP(O)(O)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Viscous liquid
Assay (unspecified)
Phosphate (as P2O5): 83.0-87.0%
Comment
May darken on storage
Impurity content
Arsenic (As): ≤30ppm

Description

Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

Solubility
Miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases. It reacts with water violently to generate heat and phosphoric acid.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text