Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Clear colorless to pale yellow to yellow to red to brown or pink
Assay (GC)
≥98.0%
Refractive Index
1.6010-1.6050 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid or viscous liquid
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.