Thermo Scientific Chemicals

Sodium dichromate dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L14890.36
500 g, Each
Thermo Scientific Chemicals

Sodium dichromate dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L14890.36
500 g, Each
Quantity
Catalog number: L14890.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-12-0
IUPAC Name
disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
Molecular Formula
Cr2H4Na2O9
InChI Key
JYDRNIYTFCBIFC-UHFFFAOYSA-N
SMILES
O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Appearance (Color)
White to yellow to orange to red
Form
Crystals or powder or crystalline powder
Assay (Iodometric Titration)
≥97.5 to ≤102.5% (non-U.S. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitor in undercoats and primers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitor in undercoats and primers.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Keep away from sources of ignition, combustible materials. Do not ingest. Do not breathe dust.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text